Synthware Flow Control Adapters with Metering Stopcock, Kemtech America
Catalog # 60000-306
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- ST Joint No.:24/40
- Cat. no.:60000-306
- Supplier No.:A732440
Specifications
About this item
Adapter has a metering stopcock between top and bottom [ST] joints to permit flow control of fluids between apparatus.
- Metering stopcock
- Top outer [ST] joint
- Bottom inner [ST] joint
Adapter features a 2 mm PTFE metering stopcock between top outer and lower inner [ST] joints. Stopcock bore is 2 mm.