Synthware Flow Control Adapters with Metering Stopcock, 90° Angle, Kemtech America
Provides on/off control with the main valve and allows delivery of preset volumes of gases or liquids to reactions using the needle valve.
- 90° angle bend
- [ST] joint
- Metering stopcock
Stopcock bore is 2 mm.
Frequently Bought Together