Synthware Vacuum/Inert Gas Double Manifold, Kemtech America
General purpose double bank manifolds are designed for use with vacuum and inert gas
- Hi vacuum valves
- Heavy wall borosilicate construction
- 4 or 5 port options available
All manifolds are supplied with 0 - 4 mm high vacuum valves for grease-free operation. Both banks are made from 22 mm medium wall tubing and are available in a variety of opening configurations. Ports are spaced 100 mm center to center and have an O.D. of 10 mm.
Frequently Bought Together