Syringes for Thin Layer Chromatography
Fixed needles, 100 µl, PTFE
Catalog # 77506-154
Supplier: INNOVATIVE LABOR SYSTEME GMBH
Supplier #: 2606702
Jump to:
Product Details & Documents
This syringe is specially designed for thin layer chromatography.
- Glass barrel is made of Borosilicate 3.3
- Precision: <±1% of the volume
The needle is developed for the needs of the e Linomat III and IV.
Spécifications
- Volume:100 µl
- Modèle:Fixed needles
- Tip Type:PTFE