Benstat™ Divider Tote Boxes, Metro™
Benstat™ Divider Tote Boxes, Metro™
Supplier:   Metropolitan Wire
Benstat™ Divider Tote Boxes, Metro™
Supplier:  Metropolitan Wire
Certificates
Benstat™ Divider Tote Boxes, Metro™
Benstat™ Divider Tote Boxes, Metro™
9 Options Available Below
Product Details & Documents
Tote boxes offer safe and efficient storage of items
Boxes, dividers, covers, and card holders are made from Benstat™, a static-dissipative polypropylene-based material with a surface resistivity of >105ohms/sq. and <1012ohms/sq. All Benstat™ products are blue.
Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.
Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes.
Product Family Options
Product Information
  • Capacité
    Max. No. of Dividers
    Interior Dimensions
    Exterior Dimensions
    Availability
    Price
  • 5.4 L (0.19 cu.ft.)
    5 long, 7 short
    16.5W×23.2L×14.9H cm (61/2×91/8×57/8")
    21W×27.6L×15.2H cm (81/4×107/8×6")
    Log in for availability
    Log in for availability
    Each
    Each - $77.41
    $77.41
    Each
       $77.41
    Supplier Number: TB91060BAS
  • 6.8 L (0.24 cu.ft.)
    7 long, 11 short
    23.5W×37.8L×8.6H cm (91/4×147/8×33/8")
    27.6W×41.9L×8.9H cm (107/8×161/2×31/2")
    Log in for availability
    Log in for availability
    Pack of 24
    Pack of 24 - $1,215.26
    $1,215.26
    Pack of 24
       $1,215.26
    Supplier Number: TB92035BAS
  • 16.7 L (0.59 cu.ft.)
    11 long, 11 short
    38.1W×38.1L×12.4H cm (15×15×47/8")
    41.9W×41.9L×12.7H cm (161/2×161/2×5")
    Log in for availability
    Log in for availability
    Each
    Each - $128.71
    $128.71
    Each
       $128.71
    Supplier Number: TB92050BAS
  • 17 L (0.60 cu.ft.)
    7 long, 11 short
    23.5W×37.8L×20H cm (91/4×147/8×77/8")
    27.6W×41.9L×20.3H cm (107/8×161/2×8")
    Log in for availability
    Log in for availability
    Each
    Each - $121.38
    $121.38
    Each
       $121.38
    Supplier Number: TB92080BAS
  • 12.5 L (0.44 cu.ft.)
    11 long, 15 short
    39.7W×52.4L×7.3H cm (155/8×205/8×27/8")
    44.5W×57.2L×7.6H cm (171/2×221/2×3")
    Log in for availability
    Log in for availability
    Each
    Each - $156.56
    $156.56
    Each
       $156.56
    Supplier Number: TB93030BAS
  • 22.4 L (0.79 cu.ft.)
    11 Long, 15 Short
    39.7W×52.4L×12.4H cm (155/8×205/8×47/8")
    44.5W×57.2L×12.7H cm (171/2×221/2×5")
    Log in for availability
    Log in for availability
    Each
    Each - $186.00
    $186.00
    Each
       $186.00
    Supplier Number: TB93050BAS
  • 27.5 L (0.97 cu.ft.)
    11 long, 15 short
    39.7W×52.4L×14.9H cm (155/8×205/8×57/8")
    44.5W×57.2L×15.2H cm (171/2×221/2×6")
    Log in for availability
    Log in for availability
    Case of 6
    Case of 6 - $1,021.20
    $1,021.20
    Case of 6
       $1,021.20
    Supplier Number: TB93060BAS
  • 37.4 L (1.32 cu.ft.)
    11 long, 15 short
    39.7W×52.4L×20H cm (155/8×205/8×77/8")
    44.5W×57.2L×20.3H cm (171/2×221/2×8")
    Log in for availability
    Log in for availability
    Each
    Each - $233.50
    $233.50
    Each
       $233.50
    Supplier Number: TB93080BAS
  • 57.5 L (2.03 cu.ft.)
    11 long, 15 short
    39.7W×52.4L×30.2H cm (155/8×205/8×117/8")
    44.5W×57.2L×30.5H (171/2×221/2×12")
    Log in for availability
    Log in for availability
    Pack of 1
    Pack of 1 - $282.45
    $282.45
    Pack of 1
       $282.45
    Supplier Number: TB93120BAS
Accessories
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-546
    82002-522
    23.2L×14H cm (91/8×51/2")
    Log in for availability
    Each - $15.41
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-552
    82002-530
    37.8L×19.1H cm (147/8×71/2")
    Log in for availability
    Each - $30.36
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Short Divider
    Short Divider
    CA14233-012
    82002-530
    23.5L×14H cm (91/4×51/2")
    Log in for availability
    Each - $19.18
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Short Divider
    Short Divider
    CA14233-004
    82002-522
    16.5L×7.6H cm (61/2×3")
    Log in for availability
    Each - $14.43
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Insert Style Cover
    Insert Style Cover
    CA14233-000
    82002-530
    Log in for availability
    Each - $76.81
Product Information
  • Catalog #
    Fits
    Availability
    Price
  • Insert Style Cover
    Insert Style Cover
    CA14233-006
    82002-522
    Log in for availability
    Each - $49.91
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-560
    82002-538; 82003-786
    53L×19.1H cm (207/8×71/2")
    Log in for availability
    Each - $33.61
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-558
    82002-538; 82003-786
    53L×14H cm (207/8×51/2")
    Log in for availability
    Each - $30.36
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Short Divider
    Short Divider
    CA14233-002
    82002-530
    23.5L×7.6H cm (91/4×3")
    Log in for availability
    Each - $16.19
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-548
    82002-530
    37.8L×7.6H cm (147/8×3")
    Log in for availability
    Each - $17.51
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-556
    82002-534, -538; 82003-786
    53L×11.4H cm (207/8×41/2")
    Log in for availability
    Each - $25.01
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-550
    82002-528, -530
    37.8L×14H cm (147/8×51/2")
    Log in for availability
    Each - $21.04
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-554
    82002-532, -534, -538; 82003-786
    53L×6.4H cm (207/8×21/2")
    Log in for availability
    Each - $25.01
Product Information
  • Catalog #
    Fits
    Exterior Dimensions
    Availability
    Price
  • Long Divider
    Long Divider
    CA82002-544
    82002-522
    23.2L×7.6H cm (91/8×3")
    Log in for availability
    Each - $14.77