VWR® Critical Swab® Swab, Foam-Over-Cotton Head
Fournisseur: VWR International
Certificats
À propos de cet article
Disposable foam-over-cotton tip swab features 100% polyurethane foam with 100 ppi and a wood shaft.
- Polyurethane foam head
- Wood shaft
Frequently Bought Together