VWR® Critical Swab® Small Foam-Over-Cotton Tipped Swab, Wooden Handle
Fournisseur: VWR International
Certificats
À propos de cet article
Small, disposable foam-over-cotton tip swab features 100% polyurethane foam with 100ppi and a wood shaft.
- Small polyurethane foam head
- Wood shaft
- Disposable
Made in the USA.
Certifications: CE marked.
Ordering information: Packaged as 20 bags of 50 swabs per bag.
Frequently Bought Together