Puritan® Small Half-Round Polyester Tipped Applicator, Acetal Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Certificates
About this item
Ideal for use as a cleaning swab for fiber optics, wafers, and disk drives
Tip is free from volatile residues, particles, contaminating adhesives, and lint. Acetal shaft. Made in the USA.
Certifications: CE marked.
Ordering information: Swabs are packaged 100 per pack, 10 packs per case.
Frequently Bought Together