Puritan® Medium Round Polyester Tipped Applicator, Polypropylene Handle, Puritan Medical Products
For use in critical environment cleaning
Chemical-resistant, nonabrasive, symmetrical tip is free from volatile residues, particles, contaminating adhesives, and lint. Polypropylene shaft. Made in the USA.
: CE marked.
: Swabs are packaged 100 per pack, 10 packs per case.
Frequently Bought Together