ContactUsLinkComponent
VWR® Critical Swab® Swab, Fine-Point Reticulated Foam Head
Catalog # 89031-294
Supplier:  VWR International
Product available on GSA Advantage®
undefined
VWR® Critical Swab® Swab, Fine-Point Reticulated Foam Head
Catalog # 89031-294
Supplier:  VWR International
Product available on GSA Advantage®
Specifications
  • Cat. no.:
    89031-294
  • Handle Length:
    161.0 mm (6.324")
  • Head Width:
    1.8 mm (0.070")
  • Head Length:
    14.6 mm (0.574")
  • Handle Width:
    2.4 mm (0.094")
Specifications
About this item
Swab features reticulated foam tip and a blue glass-filled polypropylene shaft.
  • Polypropylene shaft
  • Reticulated foam head
  • Extra rigid
For use in critical environments.