VWR® Critical Swab® Swab, Large Rectangular Foam Head
Fournisseur: VWR International
Certificats
À propos de cet article
Disposable rectangular swab features 100% polyurethane foam with 100ppi and a sturdy polypropylene handle.
- Polyurethane foam head
- Sturdy polypropylene handle
- Disposable
Thermally bonded foam head reduces contamination.
Frequently Bought Together