VWR® Critical Swab® Swab, Cone-Shaped Reticulated Foam Head
Supplier: VWR International
Certificates
About this item
Swab features reticulated foam and a blue glass-filled polypropylene shaft for extra rigidity.
- Reticulated foam
- Polypropylene shaft
- Extra rigid
For use in critical environments.
Frequently Bought Together