VWR® Critical Swab® Swab, Paddle-Shaped Foam Head
Fournisseur: VWR International
Certificats
À propos de cet article
Disposable, flexible paddle swab features 100% polyurethane foam with 100ppi and blue polypropylene shaft.
- Polyurethane foam head
- Blue polypropylene shaft
- Flexible and disposable
Frequently Bought Together