VWR® Foam-Tipped Swab
Catalog # 89009-338
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Cat. no.:89009-338
- Handle Length:149.2 mm (5.874")
- Head Width:4.6 mm (0.181")
- Head Length:16.0 mm (0.630")
- Handle Width:2.5 mm (0.098")
Specifications
About this item
Foam tip is made from soft and pliable medical-grade 100ppi polyurethane foam.
- Ideal for critical environments
- Polystyrene shaft
- Soft and pliable tip
The fine-pore, high-absorbency tip is specifically designed to retain and transfer fluids. Polystyrene shaft is scored 74.4mm (2.93") from tip.