PurSwab® Standard Foam Tipped Applicator, Polypropylene Handle, Puritan Medical Products
Supplier: Puritan Medical Products
Certificates
About this item
Lint-free, nonabrasive, shred-resistant swabs are ideal for precision cleaning
Made in the USA.
The 100ppi polyurethane foam tip is securely bonded to the polypropylene shaft.
Certifications: CE marked.
Frequently Bought Together