PurSwab® Chemical Resistant Foam Tipped Applicator, Puritan Medical Products
Fournisseur: Puritan Medical Products
Certificats
À propos de cet article
This specialty foam tipped applicator is produced with chemical resistant foam
Made in the USA.
It is slightly textured for light abrasion and will not swell or break apart with most solvents. For use in critical environments as a general cleaning swab, removing contamination and adhesives, and applying adhesives or other solutions.
Certifications: CE marked.
Ordering information: Packaged as 20 bags of 50 swabs per bag.
Frequently Bought Together