Contamination Control Swab, Foam Head, Flexible Tip, Foamtec International WCC
Catalogue # 10810-862
Fournisseur: Foamtec International
Spécifications
- Cat. no.:10810-862
- Handle Length:152.4 mm (6.000")
- Head Width:6.4 mm (0.250")
- Head Length:15.2 mm (0.600")
- Handle Width:3.2 mm (0.125")
- Supplier No.:HT1001
Spécifications
À propos de cet article
Designed to meet demanding cleanroom applications
Can be used with alcohol, Freon® 113 fluorocarbon, acetone, and other solvents for assembly cleaning, and for epoxy application. Flexible-tip swab features a 100ppi polyurethane head and a polypropylene handle.
Ordering information: Packaged in polybags.