SP Bel-Art Flaskup™ Flask Holders, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
These unique holders are perfect for keeping round and tapered bottom flasks upright.
- Polypropylene construction
- Durable and easy to clean
- Autoclavable
- Slight taper on inside wall
- Open bottom to accommodate multiple flask sizes
The inside wall has a slight taper to the open bottom, allowing a variety of flask shapes and sizes to fit into one holder.
The wide base keeps flasks stable and side slots can be used as sight windows. Molded from polypropylene, they are durable and easy to clean. Autoclavable at 121°C (250°F).
Ordering information: Assortment pack consists of three holders each of the following sizes: 10, 25, 50, and 100 mL.
Frequently Bought Together