SureSTART™ Septa, 22 mm, Level 2 High-throughput Applications
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Level 2 septa are manufactured and tested to ensure low bleeding, robustness, and reproducibility of results for your routine GC-MS and LC-MS analyses.
- Autoclavable
- Single use
- Non sterile
Choose all-purpose silicone/PTFE septa manufactured with a shore value of 50 to reduce autosampler needle issues.
SureSTART Level 2 vials have very low metal contents and leachables to ensure that they conform to the high standards you need to meet your regulatory requirements. The septa are conditioned to provide a very low level of chromatographic extractables. These products support routine analyses and deliver high quality data with an uninterrupted workflow.
Frequently Bought Together