CleanTips® Polyester Alpha® Mini Swab, Texwipe®
Product Details & Documents
The Alpha® Mini Swab is an excellent swab for cleaning small areas
The compact polypropylene handle provides support and control. Low particle generation and extractable levels. Excellent chemical resistance.
It's cleanroom laundered, double-layered, knitted Alpha® polyester head is extremely clean. Swabs feature 100% continuous-filament polyester substrate that is heat-molded (thermally-bonded) onto the handle, sealing exposed edges for reduced fiber release and eliminating the need for adhesives. Mini tip with a rigid paddle
Ordering information: Available in bag or case quantities.
Documents
Spécifications
- Désignation:Mini Polyester Swab
- Handle Length:60.0 mm (2.362")
- Overall Length:69.5 mm (2.736")
- Head Width:3.5 mm (0.138")
- Head Length:9.4 mm (0.370")
- Handle Width:2.5 mm (0.984")
- Cat. No.:TWTX754B
- Handle material:Polypropylene
- Bond:Thermal
- Head material:Knitted Alpha® Polyester
- Handle colour:Light Green