Puritan® Swab, Small Rectangular Cotton Head, Paper Shaft, Puritan Medical Products
Catalogue # CA82030-752
Fournisseur: Puritan Medical Products
Spécifications
- Cat. no.:CA82030-752
- Handle Length:146 mm (5.748")
- Head Width:4 mm (0.157")
- Head Length:4 mm (0.157")
- Handle Width:2.46 mm (0.096")
- Supplier No.:896-PC
Spécifications
À propos de cet article
For cleaning in sensitive environments.
Lint-free and absorbent single-end cotton tip. Paper shaft.