VWR® Critical Swab® Slim Cotton Double Tipped Swab, Fine Paper Handle
89031-292
Product available on GSA Advantage®
- Color:White
- Cat. no.:89031-292
- Handle Length:75.5 mm (2.972")
- Head Width:3.2 mm (0.126")
- Head Length:14.0 mm (0.551")
- Handle Width:1.5 mm (0.059")
Swab features a fine paper stem and two slim tips composed of lintless, highly absorbent cotton.
- Antistatic packing
- Lintless, highly absorbent cotton
Made in the USA.
Packaged in an antistatic leaf pack.
: CE marked.
: Packaged as 50 leaves of 25 swabs per leave.