VWR® Critical Swab® Smooth Cotton Double Tipped Swab, Paper Handle
Swab features a paper stem and two tips composed of lintless, highly absorbent cotton.
- Antistatic packing
- Lintless, highly absorbent cotton
Packaged in an antistatic leaf pack.
: CE marked.
: Packaged as 50 leaves of 25 swabs per leave.
Frequently Bought Together