VWR® Critical Swab® Swab, Cotton Head
Supplier: VWR International
Certificates
Certificates
About this item
Disposable swab with absorbent cotton tip and wood shaft.
- Autoclavable
- Disposable
Frequently Bought Together
VWR® Lab Markers
Supplier: VWR International
Acide sulfurique 0.02 N in aqueous solution analytical reagent, VWR Chemicals BDH®
Supplier: VWR International
VWR® General Purpose Laboratory Labeling Tape
Supplier: VWR International
Méthanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
Sanicult™ Hygiene Monitoring Test Kits, Starplex®
Supplier: Starplex
VWR® Laboratory Notebooks
Supplier: VWR International
VWR® Labels 'Radioactive Warning'
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
Histoplex™ Histology Containers, Starplex®
Supplier: Starplex
VWR® Stainless Steel Scalpel Blades
Supplier: VWR International
Calibration Standard, Phosphate as P, ERA
Supplier: ERA
Dibutyryl-cAMP sodium salt ≥97.0% (by HPLC)
Supplier: TCI America
SODIUM BISULFATE 35% 120 ML
Supplier: Ricca Chemical
STORAGE SOLUTION PH ELECTRODE
Supplier: Oakton
Glycol, CHEMets Visual Kit, CHEMetrics
Supplier: CHEMetrics
Whatman™ Grade GF/A Glass Microfiber Filters, Binder Free
Supplier: Cytiva
Microsep Advance Centrifugal Devices, Cytiva (Formerly Pall Lab)
Supplier: Cytiva