Rtx®-5MS GC Columns, Restek
À propos de cet article
Low-bleed columns developed for mass spectrometers and the analysis of semi-volatile pollutants.
- Low bleed
- High degree of inertness
- Internal dimensions and film thickness vary between models
The synthesis and bonding procedures for the Rtx®-5MS were modified from standard 5% phenyl polymers to address specific bleed concerns by mass spectroscopists with attention being paid to reduce bleed fragments and baseline rise
Note: The DB-5MS is a silarylene-based polymer equivalent to the Rxi-5Sil MS.
Each column is shipped with the actual test chromatograph that demonstrates the high degree of inertness for stringent environmental compounds and shows minimal baseline rise at the upper operating temperature. Also included is a test chromatogram showing the polymer bleed profile by a sensitive MS detector. Phase composition: crossbonded 5% diphenyl, 95% dimethylpolysiloxane. Temperature range: –60 to 330/350 °C (–76 to 626/662 °F).
40 Options Available Below
Product Details & Documents
Low-bleed columns developed for mass spectrometers and the analysis of semi-volatile pollutants.
- Low bleed
- High degree of inertness
- Internal dimensions and film thickness vary between models
The synthesis and bonding procedures for the Rtx®-5MS were modified from standard 5% phenyl polymers to address specific bleed concerns by mass spectroscopists with attention being paid to reduce bleed fragments and baseline rise
Note: The DB-5MS is a silarylene-based polymer equivalent to the Rxi-5Sil MS.
Each column is shipped with the actual test chromatograph that demonstrates the high degree of inertness for stringent environmental compounds and shows minimal baseline rise at the upper operating temperature. Also included is a test chromatogram showing the polymer bleed profile by a sensitive MS detector. Phase composition: crossbonded 5% diphenyl, 95% dimethylpolysiloxane. Temperature range: –60 to 330/350 °C (–76 to 626/662 °F).
Product Family Options
Informations sur le produit
- DésignationType de colonneÉpaisseur du filmØ int.LongueurAvailabilityPrice
- Rtx-5MSCapillaire0.25 µm0.32 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,125.89$1,125.89Prix promotionnel pour les nouveaux clientsEach$1,125.89Supplier Number: 12621
- Rtx-5MSCapillaire1.00 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,960.09$1,960.09Prix promotionnel pour les nouveaux clientsEach$1,960.09Supplier Number: 12653
- Rtx-5MSCapillaire1.50 µm0.53 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,666.18$1,666.18Prix promotionnel pour les nouveaux clientsEach$1,666.18Supplier Number: 12670
- Rtx-5MSCapillaire0.10 µm0.32 mm15 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.10 µm0.25 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,275.11$2,275.11Prix promotionnel pour les nouveaux clientsEach$2,275.11Supplier Number: 12611
- Rtx-5MSCapillaire0.50 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,779.62$1,779.62Prix promotionnel pour les nouveaux clientsEach$1,779.62Supplier Number: 12638
- Rtx-5MSCapillaire0.50 µm0.53 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,666.18$1,666.18Prix promotionnel pour les nouveaux clientsEach$1,666.18Supplier Number: 12640
- Rtx-5MSCapillaire1.00 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,082.58$2,082.58Prix promotionnel pour les nouveaux clientsEach$2,082.58Supplier Number: 12654
- Rtx-5MSCapillaire1.00 µm0.53 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,666.18$1,666.18Prix promotionnel pour les nouveaux clientsEach$1,666.18Supplier Number: 12655
- Rtx-5MSCapillaire1.50 µm0.53 mm15 mDiscontinued-Supplier Number: -
- Supplier Number: 12605
- Rtx-5MSCapillaire0.10 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,513.50$1,513.50Prix promotionnel pour les nouveaux clientsEach$1,513.50Supplier Number: 12609
- Rtx-5MSCapillaire0.25 µm0.25 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,203.17$1,203.17Prix promotionnel pour les nouveaux clientsEach$1,203.17Supplier Number: 12620
- Rtx-5MSCapillaire0.25 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,011.69$2,011.69Prix promotionnel pour les nouveaux clientsEach$2,011.69Supplier Number: 12623
- Rtx-5MSCapillaire0.25 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,242.89$2,242.89Prix promotionnel pour les nouveaux clientsEach$2,242.89Supplier Number: 12623-124
- Rtx-5MSCapillaire0.25 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,501.72$1,501.72Prix promotionnel pour les nouveaux clientsEach$1,501.72Supplier Number: 12624
- Rtx-5MSCapillaire0.25 µm0.32 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,439.62$2,439.62Prix promotionnel pour les nouveaux clientsEach$2,439.62Supplier Number: 12627
- Rtx-5MSCapillaire0.50 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,845.61$1,845.61Prix promotionnel pour les nouveaux clientsEach$1,845.61Supplier Number: 12639
- Rtx-5MSCapillaire1.00 µm0.25 mm15 mDiscontinued-Supplier Number: 12650
- Rtx-5MSCapillaire0.50 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,650.99$1,650.99Prix promotionnel pour les nouveaux clientsEach$1,650.99Supplier Number: 12638-127
- Rtx-5MSCapillaire0.10 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,450.82$1,450.82Prix promotionnel pour les nouveaux clientsEach$1,450.82Supplier Number: 12608
- Rtx-5MSCapillaire0.25 µm0.25 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,275.11$2,275.11Prix promotionnel pour les nouveaux clientsEach$2,275.11Supplier Number: 12626
- Rtx-5MS Agilent 6850 configurationCapillaire0.25 µm0.25 mm60 mDiscontinued-Supplier Number: -
- Supplier Number: 12623-600
- Rtx-5MSCapillaire0.25 µm0.25 mm15 mDiscontinued-Supplier Number: -
- Rtx-5MS with 10 m guard columnCapillaire0.25 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,735.20$1,735.20Prix promotionnel pour les nouveaux clientsEach$1,735.20Supplier Number: 12624-128
- Rtx-5MS with 10 m guard columnCapillaire1.00 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,650.99$1,650.99Prix promotionnel pour les nouveaux clientsEach$1,650.99Supplier Number: 12653-127
- Rtx-5MSCapillaire0.25 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,693.96$1,693.96Prix promotionnel pour les nouveaux clientsEach$1,693.96Supplier Number: 12624-125
- Rtx-5MS with 5 m guard column with presstightCapillaire0.25 µm0.25 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,028.59$2,028.59Prix promotionnel pour les nouveaux clientsEach$2,028.59Supplier Number: 12623-127
- Rtx-5MS 5" Nordion cageCapillaire0.25 µm0.32 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MS 5" Nordion cageCapillaire0.25 µm0.25 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MS with 5 m guard columnCapillaire0.25 µm0.25 mm25 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $3,781.37$3,781.37Prix promotionnel pour les nouveaux clientsEach$3,781.37Supplier Number: 12620-600
- Rtx-5MSCapillaire0.10 µm0.25 mm10 m; 5 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.10 µm0.25 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,045.91$1,045.91Prix promotionnel pour les nouveaux clientsEach$1,045.91Supplier Number: 12620-124
- Rtx-5MSCapillaire0.25 µm0.25 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,457.55$2,457.55Prix promotionnel pour les nouveaux clientsEach$2,457.55Supplier Number: 12626-124
- Rtx-5MS with 5 m guard columnCapillaire0.50 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,693.96$1,693.96Prix promotionnel pour les nouveaux clientsEach$1,693.96Supplier Number: 12639-125
- Rtx-5MSCapillaire0.50 µm0.25 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,085.16$1,085.16Prix promotionnel pour les nouveaux clientsEach$1,085.16Supplier Number: 12635-127
- Rtx-5MS with 10 m guard columnCapillaire1.00 µm0.32 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,673.16$2,673.16Prix promotionnel pour les nouveaux clientsEach$2,673.16Supplier Number: 12642-128
- Rtx-5MS Agilent 6850 configurationCapillaire0.25 µm0.25 mm15 mDiscontinued-Supplier Number: -
- Rtx-5MS Agilent 6850 configurationCapillaire0.25 µm0.25 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,496.04$2,496.04Prix promotionnel pour les nouveaux clientsEach$2,496.04Supplier Number: 12626-127
- Rtx-5MSCapillaire0.50 µm0.25 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,609.76$1,609.76Prix promotionnel pour les nouveaux clientsEach$1,609.76Supplier Number: 12638-124
- Rtx-5MS 5" Nordion cage with 5 m guard columnCapillaire0.25 µm0.25 mm30 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm20 mDiscontinued-Supplier Number: -
- Rtx-5MSCapillaire0.25 µm0.25 mm50 mDiscontinued-Supplier Number: -
- Supplier Number: 12635
- Supplier Number: 12637
- Rtx-5MSCapillaire0.50 µm0.25 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,275.11$2,275.11Prix promotionnel pour les nouveaux clientsEach$2,275.11Supplier Number: 12641
- Rtx-5MSCapillaire0.50 µm0.32 mm60 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $2,439.62$2,439.62Prix promotionnel pour les nouveaux clientsEach$2,439.62Supplier Number: 12642
- Supplier Number: 12652
- Rtx-5MSCapillaire0.25 µm0.32 mm30 mLog in pour la disponibilitéLog in pour la disponibilitéPack of 6Pack of 6 - $6,519.60$6,519.60Prix promotionnel pour les nouveaux clientsPack of 6$6,519.60Supplier Number: 12624-600
- Rtx-5MS with 5 m guard columnCapillaire1.50 µm0.53 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,147.66$1,147.66Prix promotionnel pour les nouveaux clientsEach$1,147.66Supplier Number: 12667-126
- Rtx-5MS with 5 m guard columnCapillaire1.00 µm0.53 mm15 mLog in pour la disponibilitéLog in pour la disponibilitéEachEach - $1,139.96$1,139.96Prix promotionnel pour les nouveaux clientsEach$1,139.96Supplier Number: 12652-126
- Rtx-5MS Agilent 6850 configurationCapillaire1.00 µm0.53 mm30 mDiscontinued-Supplier Number: -