Rtx®-200 GC Columns, Restek
: Restek
General-purpose columns for solvents, Freon® fluorocarbons, alcohols, ketones, silanes, and glycols.
- High-polarity column
- Different elution order than phenyl-, cyano-, and Carbowax®-containing phases
- Internal dimensions and film thickness vary between models
Can be used as a confirmation column with the Rtx-5 for phenols, nitrosamines, chlorinated pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides. Phase composition: crossbonded trifluoropropylmethylpolysiloxane modified for use with ECDs, NPDs, and MSDs.
General-purpose columns for solvents, Freon® fluorocarbons, alcohols, ketones, silanes, and glycols.
- High-polarity column
- Different elution order than phenyl-, cyano-, and Carbowax®-containing phases
- Internal dimensions and film thickness vary between models
Can be used as a confirmation column with the Rtx-5 for phenols, nitrosamines, chlorinated pesticides, chlorinated hydrocarbons, and chlorophenoxy herbicides. Phase composition: crossbonded trifluoropropylmethylpolysiloxane modified for use with ECDs, NPDs, and MSDs.
- Catalog No:
- RK15021
- RK15025
- RK15029
- RK15039
- RK15044
- RK15056
- RK15022
- RK15027
- RK15038
- RK15042
- RK15052
- RK15054
- RK15055
- RK15058
- RK15085
- RK15009
- RK15020
- RK15024
- RK15026
- RK15035
- RK15040
- RK15043
- RK15050
- RK15051
- RK15053
- RK15059
- RK15060
- RK15070
- RK15072
- RK15075
- RK15088
- RK15041
- RK15045
- RK15067
- RK15069
- RK15073
- RK15091
- RK15023
- RK15036
- RK15057
- RK15082
- 10059-566
- 10850-548
- 10850-550
- 10850-552
- 10850-554
- 10850-556
- 10850-558
- 10850-560
- Description:
- RK15021
- RK15025
- RK15029
- RK15039
- RK15044
- RK15056
- RK15022
- RK15027
- RK15038
- RK15042
- RK15052
- RK15054
- RK15055
- RK15058
- RK15085
- RK15009
- RK15020
- RK15024
- RK15026
- RK15035
- RK15040
- RK15043
- RK15050
- RK15051
- RK15053
- RK15059
- RK15060
- RK15070
- RK15072
- RK15075
- RK15088
- RK15041
- RK15045
- RK15067
- RK15069
- RK15073
- RK15091
- RK15023
- RK15036
- RK15057
- RK15082
- Temperature limit:
- –20 to 340°C
- –20 to 330°C
- –20 to 340°C
- –20 to 320°C
- –20 to 330°C
- –20 to 310°C
- –20 to 330°C
- –20 to 340°C
- –20 to 330°C
- –20 to 330°C
- –20 to 310°C
- –20 to 310°C
- –20 to 310°C
- –20 to 310°C
- –20 to 270°C
- –20 to 340°C
- –20 to 340°C
- –20 to 340°C
- –20 to 340°C
- –20 to 330°C
- –20 to 320°C
- –20 to 320°C
- –20 to 310°C
- –20 to 310°C
- –20 to 310°C
- –20 to 310°C
- –20 to 310°C
- –20 to 300°C
- –20 to 290°C
- –20 to 300°C
- –20 to 280°C
- –20 to 330°C
- –20 to 330°C
- –20 to 300°C
- –20 to 300°C
- –20 to 300°C
- –20 to 280°C
- –20 to 340°C
- –20 to 330°C
- –20 to 310°C
- –20 to 280°C
Product Family Options
- Type de colonneÉpaisseur du filmØ int.Longueur
- : 15021
Specifications
Cat. No.RK15021DescriptionRK15021Temperature limit–20 to 340°C - : 15025
Specifications
Cat. No.RK15025DescriptionRK15025Temperature limit–20 to 330°C - : 15029
Specifications
Cat. No.RK15029DescriptionRK15029Temperature limit–20 to 340°C - : 15039
Specifications
Cat. No.RK15039DescriptionRK15039Temperature limit–20 to 320°C - : 15044
Specifications
Cat. No.RK15044DescriptionRK15044Temperature limit–20 to 330°C - : 15056
Specifications
Cat. No.RK15056DescriptionRK15056Temperature limit–20 to 310°C - : 15022
Specifications
Cat. No.RK15022DescriptionRK15022Temperature limit–20 to 330°C - : 15027
Specifications
Cat. No.RK15027DescriptionRK15027Temperature limit–20 to 340°C - : 15038
Specifications
Cat. No.RK15038DescriptionRK15038Temperature limit–20 to 330°C - : 15042
Specifications
Cat. No.RK15042DescriptionRK15042Temperature limit–20 to 330°C - : 15052
Specifications
Cat. No.RK15052DescriptionRK15052Temperature limit–20 to 310°C - : 15054
Specifications
Cat. No.RK15054DescriptionRK15054Temperature limit–20 to 310°C - : 15055
Specifications
Cat. No.RK15055DescriptionRK15055Temperature limit–20 to 310°C - : 15058
Specifications
Cat. No.RK15058DescriptionRK15058Temperature limit–20 to 310°C - : 15085
Specifications
Cat. No.RK15085DescriptionRK15085Temperature limit–20 to 270°C - : 15009
Specifications
Cat. No.RK15009DescriptionRK15009Temperature limit–20 to 340°C - : 15020
Specifications
Cat. No.RK15020DescriptionRK15020Temperature limit–20 to 340°C - : 15024
Specifications
Cat. No.RK15024DescriptionRK15024Temperature limit–20 to 340°C - : 15026
Specifications
Cat. No.RK15026DescriptionRK15026Temperature limit–20 to 340°C - Capillaire0.50 µm0.25 mm15 m-: 15035
Specifications
Cat. No.RK15035DescriptionRK15035Temperature limit–20 to 330°C - : 15040
Specifications
Cat. No.RK15040DescriptionRK15040Temperature limit–20 to 320°C - Capillaire0.50 µm0.53 mm60 m-: 15043
Specifications
Cat. No.RK15043DescriptionRK15043Temperature limit–20 to 320°C - : 15050
Specifications
Cat. No.RK15050DescriptionRK15050Temperature limit–20 to 310°C - : 15051
Specifications
Cat. No.RK15051DescriptionRK15051Temperature limit–20 to 310°C - : 15053
Specifications
Cat. No.RK15053DescriptionRK15053Temperature limit–20 to 310°C - : 15059
Specifications
Cat. No.RK15059DescriptionRK15059Temperature limit–20 to 310°C - : 15060
Specifications
Cat. No.RK15060DescriptionRK15060Temperature limit–20 to 310°C - : 15070
Specifications
Cat. No.RK15070DescriptionRK15070Temperature limit–20 to 300°C - : 15072
Specifications
Cat. No.RK15072DescriptionRK15072Temperature limit–20 to 290°C - : 15075
Specifications
Cat. No.RK15075DescriptionRK15075Temperature limit–20 to 300°C - : 15088
Specifications
Cat. No.RK15088DescriptionRK15088Temperature limit–20 to 280°C - : 15041
Specifications
Cat. No.RK15041DescriptionRK15041Temperature limit–20 to 330°C - Capillaire0.50 µm0.32 mm105 m-: -
Specifications
Cat. No.RK15045DescriptionRK15045Temperature limit–20 to 330°C - Capillaire1.50 µm0.53 mm15 m-: -
Specifications
Cat. No.RK15067DescriptionRK15067Temperature limit–20 to 300°C - : 15069
Specifications
Cat. No.RK15069DescriptionRK15069Temperature limit–20 to 300°C - : 15073
Specifications
Cat. No.RK15073DescriptionRK15073Temperature limit–20 to 300°C - : 15091
Specifications
Cat. No.RK15091DescriptionRK15091Temperature limit–20 to 280°C - : 15023
Specifications
Cat. No.RK15023DescriptionRK15023Temperature limit–20 to 340°C - : 15036
Specifications
Cat. No.RK15036DescriptionRK15036Temperature limit–20 to 330°C - : 15057
Specifications
Cat. No.RK15057DescriptionRK15057Temperature limit–20 to 310°C - Capillaire3.00 µm0.53 mm15 m-: 15082
Specifications
Cat. No.RK15082DescriptionRK15082Temperature limit–20 to 280°C - Capillaire0.15 µm0.15 mm10 m-: -
Specifications
Cat. No.10059-566DescriptionTemperature limit - : 43836
Specifications
Cat. No.10850-548DescriptionTemperature limit - : 45001
Specifications
Cat. No.10850-550DescriptionTemperature limit - : 45002
Specifications
Cat. No.10850-552DescriptionTemperature limit - Capillaire0.20 µm0.18 mm40 m-: -
Specifications
Cat. No.10850-554DescriptionTemperature limit - : 45010
Specifications
Cat. No.10850-556DescriptionTemperature limit - : 45011
Specifications
Cat. No.10850-558DescriptionTemperature limit - : 45012
Specifications
Cat. No.10850-560DescriptionTemperature limit