Advanced® Bilirubin STAT-Analyzer™, Model BR2, Advanced Instruments
Analyzer provides a rapid and accurate determination of total and direct bilirubin in neonates from a single 30µL serum sample
Using spectrophotometric analysis, total bilirubin level is determined immediately and direct bilirubin level is determined in about two minutes. Analyzer is easy to use, with simple set up, calibration, and operation.
Dimensions: 330×280×180 mm (13×11×7")
: ETL-C listed. CE marked. Analyzer test system is categorized as a CLIA non-waived test.
: Manufacturer’s one-year warranty on workmanship and all parts except glass, plastic, and parts under warranty by their manufacturer.
Analyzer provides a rapid and accurate determination of total and direct bilirubin in neonates from a single 30µL serum sample
Using spectrophotometric analysis, total bilirubin level is determined immediately and direct bilirubin level is determined in about two minutes. Analyzer is easy to use, with simple set up, calibration, and operation.
Dimensions: 330×280×180 mm (13×11×7")
: ETL-C listed. CE marked. Analyzer test system is categorized as a CLIA non-waived test.
: Manufacturer’s one-year warranty on workmanship and all parts except glass, plastic, and parts under warranty by their manufacturer.
- Catalog No:
- 52197-566
- 52197-560
- CA52197-562
- Range:
- 0–30 mg/dL
- 0–30 mg/dL
- 0–30 mg/dL
- Sample size:
- 30 µL
- 30 µL
- 30 µL
- Results displayed, direct:
- 2 min.
- 2 min.
- 2 min.
- Results displayed, total:
- Instant
- Instant
- Instant
- Repeatability:
- ±0.2 mg/dL (1 S.D.)
- ±0.2 mg/dL (1 S.D.)
- ±0.2 mg/dL (1 S.D.)
- Resolution:
- 0.1 mg/dL
- 0.1 mg/dL
- 0.1 mg/dL
- Electrical:
- 100–130V, 60Hz, 20W
- 100–130V, 60Hz, 20W
- 100–130V, 60Hz, 20W
- Shipping weight:
- 5.5 kg (12 lbs.)
- 5.5 kg (12 lbs.)
- 5.5 kg (12 lbs.)
Product Family Options
- Désignation
- : B2A101
Specifications
Cat. No.52197-566Range0–30 mg/dLSample size30 µLResults displayed, direct2 min.Results displayed, totalInstantRepeatability±0.2 mg/dL (1 S.D.)Resolution0.1 mg/dLElectrical100–130V, 60Hz, 20WShipping weight5.5 kg (12 lbs.) - : B2A102
Specifications
Cat. No.52197-560Range0–30 mg/dLSample size30 µLResults displayed, direct2 min.Results displayed, totalInstantRepeatability±0.2 mg/dL (1 S.D.)Resolution0.1 mg/dLElectrical100–130V, 60Hz, 20WShipping weight5.5 kg (12 lbs.) - : BR2
Specifications
Cat. No.CA52197-562Range0–30 mg/dLSample size30 µLResults displayed, direct2 min.Results displayed, totalInstantRepeatability±0.2 mg/dL (1 S.D.)Resolution0.1 mg/dLElectrical100–130V, 60Hz, 20WShipping weight5.5 kg (12 lbs.)