Automatic Liquid Handling Syringes, H-XL
Supplier: INNOVATIVE LABOR SYSTEME GMBH
Jump to:
À propos de cet article
XL series syringes are designed to work with most industry standard pump designs e.g. Tecan Cavro.
- Luer or ¼-28 threaded end caps
- Broad variety of syringe interfaces
- Seals available in PTFE, UHMWPE, or custom UHMWPE blends
- Customized designts and materials to suit application requirements
- Stroke 60 mm
Glass barrel: Borosilicate 3.3
Front fitting: Glass barrel: Borosilicate 3.3
15 Options Available Below
Product Details & Documents
XL series syringes are designed to work with most industry standard pump designs e.g. Tecan Cavro.
- Luer or ¼-28 threaded end caps
- Broad variety of syringe interfaces
- Seals available in PTFE, UHMWPE, or custom UHMWPE blends
- Customized designts and materials to suit application requirements
- Stroke 60 mm
Glass barrel: Borosilicate 3.3
Front fitting: Glass barrel: Borosilicate 3.3
Product Family Options
PTFE-seals
Informations sur le produit
- VolumeAvailabilityPrice
- Supplier Number: 2607511Spécifications:
More
Specifications
Cat. No.77506-220 - Supplier Number: 2607711Spécifications:
More
Specifications
Cat. No.77506-238 - Supplier Number: 2607811Spécifications:
More
Specifications
Cat. No.77506-252 - Supplier Number: 2607911Spécifications:
More
Specifications
Cat. No.77506-264 - Supplier Number: 2607013Spécifications:
More
Specifications
Cat. No.77506-158 - Supplier Number: 2607033Spécifications:
More
Specifications
Cat. No.77506-176 - Supplier Number: 2607053Spécifications:
More
Specifications
Cat. No.77506-190 - Supplier Number: 2607073Spécifications:
More
Specifications
Cat. No.77506-200
PTFE-seals with silicone
Informations sur le produit
- VolumeAvailabilityPrice
- Supplier Number: 2645017Spécifications:
More
Specifications
Cat. No.77506-402 - Supplier Number: 2645037Spécifications:
More
Specifications
Cat. No.77506-404 - Supplier Number: 2645077Spécifications:
More
Specifications
Cat. No.77506-410
HDPE-seals
Informations sur le produit
- VolumeAvailabilityPrice
- Supplier Number: 2645918Spécifications:
More
Specifications
Cat. No.77506-414 - Supplier Number: 2645038Spécifications:
More
Specifications
Cat. No.77506-406 - Supplier Number: 2645058Spécifications:
More
Specifications
Cat. No.77506-408 - Supplier Number: 2645078Spécifications:
More
Specifications
Cat. No.77506-412