PYREX® Vacuum Traps, Corning
PYREX® Vacuum Traps, Corning
   Sustainable Options Available
Supplier:   Corning
PYREX® Vacuum Traps, Corning
Supplier:  Corning
   Sustainable Options Available
Certificates
PYREX® Vacuum Traps, Corning
6 Options Available Below
Product Details & Documents
These cold traps are made of PYREX® borosilicate glass, clear [ST] joints on both outer and inner joints. The top and side tubulations are the same Ext.Ø. as the centre tube and have a length of 75 mm.
Specifications for Product Family
Specifications
  • Catalog No:
  • 89091-280
  • 89091-338
  • 89091-348
  • 89091-382
  • 55096-188
  • 89091-468
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Specifications
Product Family Options
Product Information
  • Longueur
    ST Joint No.
    Ø ext.
    Inner Tube O.D.×L
    Availability
    Price
  • 200 mm
    29/42
    28 mm
    10×175 mm
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,620.00
    $1,620.00
    Case of 4
       $1,620.00
    Supplier Number: 7729-28
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-280
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 225 mm
    34/45
    32 mm
    13×200 mm
    Log in for availability
    Log in for availability
    Case of 4
    Case of 4 - $1,840.00
    $1,840.00
    Case of 4
       $1,840.00
    Supplier Number: 7729-32
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-338
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 mm
    40/50
    38 mm
    16×225 mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $1,090.00
    $1,090.00
    Case of 2
       $1,090.00
    Supplier Number: 7729-38
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-348
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 mm
    45/50
    42 mm
    20×225 mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $1,210.00
    $1,210.00
    Case of 2
       $1,210.00
    Supplier Number: 7729-41
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-382
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 mm
    50/50
    48 mm
    20×225 mm
    Log in for availability
    Log in for availability
    Case of 1
    Case of 1 - $728.00
    $728.00
    Case of 1
       $728.00
    Supplier Number: 7729-48
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    55096-188
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 250 mm
    55/50
    5 mm
    20×225 mm
    Log in for availability
    Log in for availability
    Case of 2
    Case of 2 - $1,340.00
    $1,340.00
    Case of 2
       $1,340.00
    Supplier Number: 7729-51
    Specifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-468
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact