Synthware Two-Neck Reaction Flask
Fournisseur: Kemtech America
Certificats
À propos de cet article
Reaction flasks are supplied with a 2 mm glass stopcock that functions as a constant pressure retention device.
- Heavy wall borosilicate construction
- Ground glass joint
- All glass stopcock
- Angled side neck
- Sidearm with stopper
With a 2 mm glass stopcock having a constant pressure retention device.
Frequently Bought Together