Synthware Reaction Flask with Glass Stopcock, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Reaction flasks are supplied with a 2 mm glass stopcock.
- Heavy wall borosilicate construction
- Ground glass joint
- All glass stopcock
Vessels feature a spring retention device for smooth turning purposes. Accessories and replacement retention kits are sold separately.
Frequently Bought Together