Synthware Reaction Tube Flask with High Vacuum Valve, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Reaction tube flasks are equipped with a high vacuum valve that features a PTFE O-ring at the tip and a PTFE wiper to protect the Viton® O-ring on the shaft.
- Heavy wall borosilicate construction
- Ground glass joint
- Teflon® covered high vacuum valve included
Valve size: 0 to 4 mm.
Frequently Bought Together