ContactUsLinkComponent
T-Shaped Plastic Spreaders, Sterile
T-Shaped Plastic Spreaders, Sterile
Catalog # 30002-115
Supplier:  Copan Diagnostics
undefined
T-Shaped Plastic Spreaders, Sterile
Catalog # 30002-115
Supplier:  Copan Diagnostics
Supplier Number:  COPTS-10
Specifications
  • Cat. no.:
    30002-115
  • Packaging:
    10/Peel pack
Specifications
About this item
Designed for spreading and dispersing liquids across the surface of agar culture plates.
Spreaders are used for performing bacterial CFU plate counts on water, milk, urine, and other liquid samples. Smooth rounded surfaces prevent cutting or gouging of agar during inoculation of culture plates. Design allows for even pressure to be applied across the entire length of the T-bar in contact with the agar surface.