Synthware Modified Schlenk Tube Flasks
Supplier: Kemtech America
Jump to:
À propos de cet article
Flasks are available with a choice of either 2 mm glass stopcock, 2 mm PTFE stopcock, or 0 to 4 mm high vacuum valve.
- Heavy wall borosilicate construction
- Ground glass joint
- 3 stopper options
Our modified schlenk tube are available with Hi-Vacuum Valves, glass, or Teflon® stopcocks.
37 Options Available Below
Product Details & Documents
Flasks are available with a choice of either 2 mm glass stopcock, 2 mm PTFE stopcock, or 0 to 4 mm high vacuum valve.
- Heavy wall borosilicate construction
- Ground glass joint
- 3 stopper options
Our modified schlenk tube are available with Hi-Vacuum Valves, glass, or Teflon® stopcocks.
Product Family Options
Informations sur le produit
- CapacitéDésignationST Joint SizeAvailabilityPrice
- Supplier Number: F901410
- Supplier Number: F901425
- Supplier Number: F901450
- Supplier Number: F901950
- Supplier Number: F902450
- Supplier Number: F901100
- Supplier Number: F909100
- Supplier Number: F904100
- Supplier Number: F901200
- Supplier Number: F909200
- Supplier Number: F904200
- Supplier Number: F904500
- Supplier Number: F904000
- Supplier Number: F901410G
- Supplier Number: F901425G
- Supplier Number: F901450G
- Supplier Number: F901950G
- Supplier Number: F902450G
- Supplier Number: F901100G
- Supplier Number: F909100G
- Supplier Number: F904100G
- Supplier Number: F901200G
- Supplier Number: F904200G
- Supplier Number: F904500G
- Supplier Number: F904000G
- Supplier Number: F901425H
- Supplier Number: F901450H
- Supplier Number: F901950H
- Supplier Number: F902450H
- Supplier Number: F901100H
- Supplier Number: F909100H
- Supplier Number: F904100H
- Supplier Number: F901200H
- Supplier Number: F909200H
- Supplier Number: F904200H
- Supplier Number: F904500H
- Supplier Number: F904000H
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm glass plug
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement 2 mm PTFE Plug
Informations sur le produit
- Catalog #I.D.×WO-Ring SizeAvailabilityPrice
- Viton® O-Ring
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Sstainless steel retention for glass stopcock, 2 mm (12/30 ST joint size)Replacement Sstainless steel retention for glass stopcock, 2 mm (12/30 ST joint size)60005-186Log in pour la disponibilitéEach - $5.81