VWR® Capsules, Porcelain
Chemical-porcelain capsules are glazed inside and out, except for rim.
- Available in flat-form and tall-form designs
- May feature either a flat or rounded bottom
- Straight sides
Flat-form capsule 89037-954 features straight sides and a flat bottom. Tall-form combustion capsules feature a rounded bottom.
Frequently Bought Together