ContactUsLinkComponent
HardyDisk™ Rezafungin, 5 µg, (RZF5)
Catalog # CA77497-274
CAS Number:  
undefined
HardyDisk™ Rezafungin, 5 µg, (RZF5)
Catalog # CA77497-274
Supplier Number:  Z9471
CAS Number:  
Specifications
  • Pack type:
    1×50 disks
  • Description:
    Rezafungin, (RZF5)
  • Size:
    5 µg
  • Cat. no.:
    CA77497-274
Specifications
About this item
Rezafungin is a new antifungal agent that combats invasive fungal infections. This marks the first new treatment option approved for individuals with candidemia and invasive candidiasis in over ten years.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.