ContactUsLinkComponent
HardyDisk™ AST Imipenem/Relebactam, IMR10/25
  CA77497-268
 :  
undefined
HardyDisk™ AST Imipenem/Relebactam, IMR10/25
  CA77497-268
 :  Z9441
 :  
  • Pack type:
    1×50 disks
  • Description:
    HardyDisk Imipenem/Relebactam, IMR10/25
  • Cat. no.:
    CA77497-268
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
  • Hardy Diagnostics offers a complete line of all commonly used antibiotics
  • Compatible with the BBL dispensers
  • Improved cartridge design helps to prevent jamming and tearing
  • HardyDisk™ AST cartridges are packaged in an all new canister that features a desiccant built into the lid
  • Kirby-Bauer accessories available: Mueller Hinton Agar, McFarland Standard, calipers, and much more
  • The last disk in each cartridge is clearly marked with dark lines around its edge to signify the need for refill
 : Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.