HardyDisk™ AST Cephalothin, CF30
Catalog # CA77497-266
Supplier: HARDY DIAGNOSTICS CA
CAS Number:
Specifications
- Pack type:5×50 disks
- Description:HardyDisk Cephalothin, CF30
- Cat. no.:CA77497-266
Specifications
About this item
HardyDisk™ AST's are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST) also known as disk diffusion or Kirby-Bauer testing.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.