Synthware Distillation Receiving Sets, Distribution Adapter, Kemtech America
Fournisseur: Kemtech America
Certificats
À propos de cet article
Rotating cow receiver allows four flasks to be progressively filled.
- Heavy wall Borosilicate construction
- Ground glass joint
- Keck clips included
The distillation receiver set allows you to distill into three separate flasks.
Ordering information: Set components are sold separately.
Frequently Bought Together