Accessories
PTFE plug, 4 mm
Catalog # 60005-116
Supplier: Kemtech America
Supplier #: S181530
Jump to:
Product Details & Documents
Replacement Teflon® stopcock.
- 1:5 taper
- Straight bore
- O-ring included
Straight bore, Teflon®, stopcock is a direct replacement for a number of Kemtech products.
Specifications
- Désignation:PTFE plug, 4 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Replacement 2 mm PTFE Plug