Sol-M® Luer Adapter | Avantor
Sol-M® Luer Adapter
Luer adapter, 20G
Catalog # 77528-890
Fournisseur #:  LA21920
Sol-M® Luer Adapter
Catalog # 77528-890
Sol-M® Luer Adapter
Sol-M® Luer Adapter
Luer adapter, 20G
Price
$46.73
Log in pour la disponibilité
Pack of 100 Case of 1000
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The Sol-M® luer adapter is designed to work in a variety of blood collection applications by securely connecting to female Luer connections.
  • Blue cap to differentiate from other blood collection devices
  • Designed to ensure secure connections
An ideal accessory to use with blood collection sets that feature a holder for evacuated tubes. Fitted with rubber sleeve to ensure no blood spills or exposure.
Spécifications
  • Désignation:
    Luer adapter, 20G