AssureWipe™ Optical Wipes
Medicom AssureWipe™ optical wipes, White, Non sterile, 11.4×20.95 cm (4.5×8.25")
Catalog # CA77497-248
Supplier: Medicom
Supplier #: OW40045
Jump to:
Product Details & Documents
Optical wipes intended for gentle cleaning of optical glass and lenses, preventing scratches and maintaining clarity.
- Single-ply specialty/delicate task wiper
- Ideal for optical glass and lenses
- Built-in polyfilm dust layer, delicate duty cleaning, optimal dispensing mechanism
- Non-sterile and latex-free
Store in a cool dry place. Application in industrial, laboratory and research, medical fields.
Packaging: Pack of 60 boxes/case; 280 wipes/box.
Specifications
- Sterility:Non sterile
- Color:White
- Désignation:Medicom AssureWipe™ optical wipes
- W×L:11.4×20.95 cm (4.5×8.25")