Premium Wrist Support, OK-1® Safety, OccuNomix
Fournisseur: Occunomix International, LLC
Certificats
À propos de cet article
The wrist support features variable stretch elastic, putting the strongest part of the support on the wrist area where it is needed, while taking some pressure off the hand and forearm for greater comfort. It helps to keep the wrist in a neutral position. The unique closure allows the support to be tightened by pulling across the back of the hand.
- An 8 cm (3") wide, wrap-around strap also adds support on the wrist area.
- Constructed of sturdy neoprene and elastic, the wrist support includes soft terry material and soft nylon against the skin for comfor
- closure helps limit wrist extension (upward motion of the hand) and wrist flexion (downward motion of the hand), while allowing full dexerity of the fingers and thumb
- Available in two sizes
X-Small: 11 - 14 cm (4.5 - 5.5") wrist circumference
Small: 14 - 17 cm (5.5 - 6.5") wrist circumference
Ordering information: Measure wrist circumference and order applicable size. Must specify left or right hand when ordering.
Frequently Bought Together