Accessories for Synthware Friedrichs Condensers with Removable Hose Connections
Supplier: Kemtech America
Certificates
About this item
Caps and removable hose connection for use with GL thread.
- Nylon construction
- GL thread
- Caps and removable hose connection
- Caps and hose connection sold separately
Removable hose connections or made of durable nylon, but can be easily cleaned and replaced when needed.
Frequently Bought Together