Disposable Plastic Restraints, AIMS™
À propos de cet article
Rat/mouse restraints are ideal for tail tattooing, tail vein injections, lp injections, oral dosing, decapitation, and general restraint
Humane, safe, and disposable. Cone-shaped bags provide easier animal capture and increased animal comfort.
19 Options Available Below
Product Details & Documents
Rat/mouse restraints are ideal for tail tattooing, tail vein injections, lp injections, oral dosing, decapitation, and general restraint
Humane, safe, and disposable. Cone-shaped bags provide easier animal capture and increased animal comfort.
Product Family Options
Informations sur le produit
- DésignationColorTailleAvailabilityPrice
- Supplier Number: 1801CV
- Supplier Number: 1802CV
- Supplier Number: 1803CV
- Supplier Number: 1801BV
- Supplier Number: 1802BV
- Supplier Number: 1803BV
- Supplier Number: 1801RV
- Supplier Number: 1802RV
- Supplier Number: 1803RV
- Supplier Number: 1800C
- Supplier Number: 1801C
- Supplier Number: 1802C
- Supplier Number: 1803C
- Supplier Number: 1801B
- Supplier Number: 1802B
- Supplier Number: 1803B
- Supplier Number: 1801R
- Supplier Number: 1802R
- Supplier Number: 1803R