Max® and Max Lite® Earplugs, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Pre-shaped foam earplugs feature a smooth outer skin for maximum user comfort.
- Disposable, non-irritating and non-allergenic
- Available with or without connecting cord
- Max Lite® low-pressure self-adjusting foam fits smaller ear canals
Certifications: Meet ANSI S3.19 standard. CE marked. CSA certified.
Ordering information: Earplugs 55533-026 are bulk-packed for use in the Howard Leight dispenser (see 89013-668 series).
Frequently Bought Together