Vee Gee Micro Volumetric Flasks
Supplier:   Heathrow Scientific
Vee Gee Micro Volumetric Flasks
Certificates
Vee Gee Micro Volumetric Flasks
Vee Gee Micro Volumetric Flasks
2 Options Available Below
Product Details & Documents
These micro volumetric flasks feature machine-blown bodies and heavy-beaded, heavy-tubing necks that are tooled for standard taper glass stoppers.
  • Made of premium borosilicate glass with a low coefficient of expansion
  • Designed 'to contain' with a temperature of calibration at 20 °C (68 °F)
  • The graduation line is sharply defined and of uniform width
  • Each flask has a large marking square and includes a glass stopper
  • Features a hexagonal base for added stability
Our flasks are constructed of premium borosilicate glass, ensuring exceptional clarity and resistance to chemical interactions. Each volumetric flask is supplied with a standard taper glass stopper which provides a leakproof seal. This airtight seal not only preserves the integrity of your samples but also enhances the reliability of your volumetric measurements.
 : Meets ASTM E-288 Class A specifications and USP standards for glassware.
Product Family Options
Product Information
  • Capacité
    ST Stopper No.
    Tolerance
    Availability
    Price
  • 1 ml
    #9
    ±0.01 ml
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $151.47
    $151.47
    New Customer Promotional Price
    Pack of 6
       $151.47
    Supplier Number: 2306A-1
  • 2 ml
    #9
    ±0.015 ml
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 6
    Pack of 6 - $158.54
    $158.54
    New Customer Promotional Price
    Log in for availability
    Case of 576
    Case of 576 - $15,219.46
    $15,219.46
    New Customer Promotional Price
    Pack of 6
       $158.54
    Case of 576
       $15,219.46
    Supplier Number: 2306A-2