Foreline Trap, Edwards | Avantor
Foreline Trap, Edwards
Foreline Trap
Catalog # CAA133-05-000
Supplier:   Edwards
Fournisseur #:  A13305000
Foreline Trap, Edwards
Catalog # CAA133-05-000
Supplier:  Edwards
Foreline Trap, Edwards
Foreline Trap, Edwards
Foreline Trap
Price
$1,910.32
Log in pour la disponibilité
Each
Product Details & Documents
For clean pumping systems to prevent back-migration of rotary pump oil vapor into the system
Note: To minimize vapor absorption during pump down, a valved bypass line is recommended.
It is ideal for two-stage pumps operating below 1mbar. Trap is filled with activated alumina, which has excellent oil vapor trapping properties. For models RV3, RV5, RV8, and R12.
Spécifications
  • Désignation:
    Foreline Trap