Soil/Sediment Sifter
Soil/sediment sifter
  470356-314
 :  SST-200
Soil/Sediment Sifter
  470356-314
Soil/Sediment Sifter
Soil/Sediment Sifter
Soil/sediment sifter
$98.05
Each
Cut through the clutter with this soil/sediment sifter. This handy tool is the real deal for serious geology work and earth science exploration.
  • 2 mm mesh
  • Overall dimensions: 61×41 cm (Length×Width)
  • Height: 6 cm without handle, 8 cm with handle
  • Ideal for soil and general particle sifting
  • Handles for easy use
Count on this sifter to deliver precision sorting of soil, sediment, and other fine particles. The sturdy wood frame and rust-resistant steel mesh are built to withstand the demands of classroom use and outdoor fieldwork.
Whether you're a teacher looking to equip your science lab or an avid amateur geologist, this 24×16×3" sifter is a must-have for accurately separating and analyzing materials.
  • Désignation:
    Soil/sediment sifter