Portable Digital Field Scale
Portable digital field scale, 600 g
470356-296
: FIELD-SCALE
This remarkable little scale is sturdy and easy to use.
- Six units: g, oz., gn, ozt, dwt, ct
- Two easy to use buttons
- LCD with blacklight
- Easy calibration
A good digital scale is indipensable, both in the field and collection room.
Accuracy: ±0.1
- Désignation:Portable digital field scale
- Weighing Capacity:600 g